HEK293/Human CCR8 Stable Cell Line 1 * 2 Vial
Catalog # ACRBCHEK-ATP140-2
Supplier: ACRO Biosystems
Frequently Bought Together
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Supplier: VWR Chemicals
Visual colour kits, iron, MQuant®, Supelco®
Supplier: Merck
Sodium hypochlorite (12% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
UltraClear® for histology, J.T.Baker®
Supplier: Avantor
Test strips, sulphite, MQuant®, Supelco®
Supplier: Merck
Test kits, photometric, iron, LCK cuvette tests
Supplier: Hach
Test kits, photometric, ammonium, LCK cuvette tests
Supplier: Hach
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
Test strips, total hardness, MQuant®, Supelco®
Supplier: Merck
VWR® Cryolab Q Path, Freezing Aerosol
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN
Supplier: Hach