ContactUsLinkComponent
PE-Labeled Rhesus macaque Mamu-A 01,B2M Tetramer Protein MAM-RP2H4 is expressed from human 293 cells HEK293. It contains AA Gly 25 - Pro 307 Mamu-A 01, Ile 21 - Met 119 B2M Accession Q5TM76 Mamu-A 01, Q6V7J5 B2M. 1 * 200 µG
Catalog # ACRBMAM-RP2H4-200
Supplier:  ACROBIOSYSTEMS
PE-Labeled Rhesus macaque Mamu-A 01,B2M Tetramer Protein MAM-RP2H4 is expressed from human 293 cells HEK293. It contains AA Gly 25 - Pro 307 Mamu-A 01, Ile 21 - Met 119 B2M Accession Q5TM76 Mamu-A 01, Q6V7J5 B2M. 1 * 200 µG
Catalog # ACRBMAM-RP2H4-200
Supplier:  ACROBIOSYSTEMS