ContactUsLinkComponent
Adalimumab is a human monoclonal antibody against TNF-alpha. It is produced by recombinant D technology using a mammalian cell expre ssion system. It consists of 1330 amino acids and has a molecular weight of approximately 148 kilodaltons. 1 * 200 µG
Catalog # ACRBADB-BY17C-200
Supplier:  ACROBIOSYSTEMS
Adalimumab is a human monoclonal antibody against TNF-alpha. It is produced by recombinant D technology using a mammalian cell expre ssion system. It consists of 1330 amino acids and has a molecular weight of approximately 148 kilodaltons. 1 * 200 µG
Catalog # ACRBADB-BY17C-200
Supplier:  ACROBIOSYSTEMS