ContactUsLinkComponent
Biotinylated Human HLA-A 2402,B2M Monomer Protein HLM-H82W4 is expressed from human 293 cells HEK293. It contains AA Gly 25 - Thr 30 5 HLA-A 2402, Ile 21 - Met 119 B2M Accession AAA59600.1 HLA-A 2402, P61769 B2M. 1 * 25 µG
Catalog # ACRBHLM-H82W4-25UG
Supplier:  ACROBIOSYSTEMS
Biotinylated Human HLA-A 2402,B2M Monomer Protein HLM-H82W4 is expressed from human 293 cells HEK293. It contains AA Gly 25 - Thr 30 5 HLA-A 2402, Ile 21 - Met 119 B2M Accession AAA59600.1 HLA-A 2402, P61769 B2M. 1 * 25 µG
Catalog # ACRBHLM-H82W4-25UG
Supplier:  ACROBIOSYSTEMS