ContactUsLinkComponent
Biotinylated Human HLA-C 070201,B2M Monomer Protein HLM-H82Ed is expressed from human 293 cells HEK293. It contains AA Cys 25 - Ile 308 HLA-C 070201, Ile 21 - Met 119 B2M Accession P10321-1 HLA-C 070201, P61769-1 B2M. 1 * 25 µG
Catalog # ACRBHLM-H82ED-25UG
Supplier:  ACROBIOSYSTEMS
Biotinylated Human HLA-C 070201,B2M Monomer Protein HLM-H82Ed is expressed from human 293 cells HEK293. It contains AA Cys 25 - Ile 308 HLA-C 070201, Ile 21 - Met 119 B2M Accession P10321-1 HLA-C 070201, P61769-1 B2M. 1 * 25 µG
Catalog # ACRBHLM-H82ED-25UG
Supplier:  ACROBIOSYSTEMS