Orders LinkContactUsLinkComponent
Bacillol® Zero Tissues
Bacillol® zero tissues XL, 80 tissues flowpack, 200×300 mm
  BODE9819360
Bacillol® Zero Tissues
  BODE9819360
Bacillol® Zero Tissues
Bacillol® zero tissues XL, 80 tissues flowpack, 200×300 mm
18,30 €
1 * 1 ST
Organic-acid based, extensively effective and material friendly cleaning and disinfecting wipes.
  • 100% plastic free nonwoven
  • Virucidal in 2 minutes
  • Free from alcohol, QAC, oxidants, perfume and dye
  • >98,5% natural origin ingredients
  • Patented formulation - organic acid complex
  • No hazardous labeling - useable without gloves
  • Excellent material compatibility with all surfaces close to the patient
  • Description:
    Bacillol® zero tissues XL, 80 tissues flowpack, 200×300 mm
  • Seq:
    01