Bacillol® Zero Tissues
Bacillol® zero tissues XL, 200×300 mm
Catalog # BODE9819360_1
Supplier: BODE CHEMIE
Jump to:
Product Details & Documents
Organic-acid based, extensively effective and material friendly cleaning and disinfecting wipes.
- 100% plastic free nonwoven
- Virucidal in 2 minutes
- Free from alcohol, QAC, oxidants, perfume and dye
- >98,5% natural origin ingredients
- Patented formulation - organic acid complex
- No hazardous labeling - useable without gloves
- Excellent material compatibility with all surfaces close to the patient
Specifications
- Description:Bacillol® zero tissues XL, 200×300 mm
- Seq:02



