Order Entry
Germany
Orders LinkContactUsLinkComponent
Mouse Recombinant IL-19

Mouse Recombinant IL-19

50 µG
Catalog # STMC78207.1
Marketsource
Mouse Recombinant IL-19
Catalog # STMC78207.1
Marketsource
CAS Number:  
Mouse Recombinant IL-19
Mouse Recombinant IL-19

50 µG

CAS Number:  
Price
762,00 €
Log in for availability
1 * 50 µG
Product Details & Documents

Interleukin 19 (IL-19) is a member of the IL-10 cytokine family and is produced by keratinocytes, B cells, and monocytes (Romer et al.; Wolk et al.). Expression of IL-19 can be induced by granulocyte-macrophage colony-stimulating factor (GM-CSF) or lipopolysaccharide (LPS; Gallagher et al.). IL-19 is considered to be a proinflammatory cytokine, as it upregulates IL-6 and tumor necrosis factor alpha (TNF-α; Liao et al. 2002). IL-19 binds the IL-20 receptor complex (IL-20R) which comprises IL-20R alpha and IL-20R beta to activate the STAT3 pathway (Dumoutier et al.). IL-19 also induces T-helper cell differentiation towards a Th2 response, resulting in the production of IL-10 and additional IL-19 (Liao et al. 2002; Liao et al. 2004). IL-19 has been implicated in aging, vascular disease, Type I diabetes, and rheumatoid arthritis.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    50 µG
  • Cat. No.:
    STMC78207.1
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    IL-19
  • Purity:
    ≥95%
  • Species:
    Mouse

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.