Erlenmeyer Flasks with screw cap
250 ml, GL 45
Catalog # NEUB0748-10-025045
Supplier: NEUBERT VOLUME GLASSWAERE
Jump to:
Product Details & Documents
Borosilicate glass 3.3, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Volume graduations and labelling area. Screw cap with PTFE coated seal.
Specifications
- Thread:GL 45
- Capacity:250 ml
- Seq:01