Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
COMPONENTS 5 500MG/L EACH OF 1 * 50 mL
Catalog # CMPLRD108879L05.20
Supplier:
CUSTOM MADE CHEMICALS LAB
Expand image
COMPONENTS 5 500MG/L EACH OF 1 * 50 mL
Catalog # CMPLRD108879L05.20
Supplier:
CUSTOM MADE CHEMICALS LAB