0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
RECOMBINANT HUMAN IL-1Β (CHO) 1 * 10 µG
INVVRCYC-HIL1B
:
Invivogen
RECOMBINANT HUMAN IL-1Β (CHO) 1 * 10 µG
INVVRCYC-HIL1B
:
Invivogen