CD3Ε RECOMBINANT MONOCLONAL ANTIBODY (145-2C11) (HAMSTER IGGΚ) 1 * 200 µG
Artikelnr. ENZOENZABS4250200
Supplier: Enzo Life Sciences
CD3Ε RECOMBINANT MONOCLONAL ANTIBODY (145-2C11) (HAMSTER IGGΚ) 1 * 200 µG
Artikelnr. ENZOENZABS4250200
Supplier: Enzo Life Sciences
Frequently Bought Together
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Cell culture flasks, Nunc™
Supplier: Thermo Fisher Scientific
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection