ANTIGEN RETRIEVAL REAGENT, PH 9 (10X) 1 * 100 mL
ENZOENZACC1130100
Frequently Bought Together
[EN]MARCING DYE BLUE 1 * 59 mL
: BRADLEY
Sodium hydroxide 50% in aqueous solution, TECHNICAL
: VWR Chemicals
HAEMATOXYLIN GILL 1 3800ML 1 * 1 items
: Epredia
Fast Red RC Salt, Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
Bromocresol green-methyl red solution mixed indicator, Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
pH Buffer solutions, CX-1
: Thermo Fisher Scientific