IL-3 (MOUSE), (RECOMBINANT) 1 * 100 µG
Catalog # ENZOALX2018210100
Supplier: Enzo Life Sciences
Frequently Bought Together
VWR®, Microcentrifuge Tubes and Screw Caps
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
Petri dishes, square, Corning® Gosselin™
Supplier: Corning
Ice pans and buckets, multi-purpose, Corning®
Supplier: Corning
Cryoboxes, 133×133 mm
Supplier: TENAK
VWR®, Reagent Reservoir, 175 ml
Supplier: VWR Collection
VWR®, Microcentrifuge Tubes, Low-Binding
Supplier: VWR Collection
VWR® Cellulose Stoppers
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Serological pipettes, Stripette™
Supplier: Corning
Corning® Treated Culture Dishes
Supplier: Corning
Individually Wrapped Serological Pipettes, Falcon®
Supplier: Corning
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
Supplier: VWR Collection
Cell-strainer, poriegrootte 100µm, geel 1 * 600 items
Supplier: SARSTEDT
Lids for microplates, Matrix
Supplier: THERMO MATRIX TECHNOLOGIES