STEM CELL FACTOR (RAT), (RECOMBINANT) 1 * 10 µG
ENZOALX2018060010
Frequently Bought Together
VWR®, Square Bottles, PET
: LAB3 OWN PRODUCTION
Hb 301 microcuvettes 1 * 200 ST
: MARKET SOURCE PART PROCESS
Tube 1,5ml with white screw cap non-skirted sterile 1 * 2.500 items
: Greiner Bio-One
Transfer pipettes
: SARSTEDT
Cryogenic vials, CryoELITE®, WHEATON®
: DWK Life Sciences
Racks and lids for storage 2D barcoded tubes, Matrix™
: THERMO MATRIX TECHNOLOGIES
BOTTLE PET 250ML SQUARE T/E GREEN CAP 1 * 200 items
: LAB3 OWN PRODUCTION
Corning® Gosselin™ Petri Dish, Aseptic
: Corning
2,0ml Reinforced Tubes mit Silikon O-Rin 1 * 1.000 ST
: MARKET SOURCE PART PROCESS
OmniTop Sample Tubes® Exact Volume Sampling System (EVSS)
: Avantor Fluid Handling
Microcentrifuge tubes, 1,5 ml
: KARTELL