Human recombinant Hsp 60
200 µG
Catalog # ENZOADISPP540200
Supplier: Enzo Life Sciences
Jump to:
Product Details & Documents
Produced in E. coli.
Specifications
Specifications
- Pk:200 µG
- Cat. No.:ENZOADISPP540200
- Protein/peptide type:Recombinant
- Purification:-
- Protein/peptide name:Hsp 60
- Formulation:Liquid
- Purity:-
- Species:Human



