Order Entry
Germany
Orders LinkContactUsLinkComponent
Human recombinant Hsp 60

Human recombinant Hsp 60

50 µG
Catalog # ENZOADISPP540050
Supplier:   Enzo Life Sciences
Human recombinant Hsp 60
Catalog # ENZOADISPP540050
Supplier:  Enzo Life Sciences
CAS Number:  
Human recombinant Hsp 60

50 µG

CAS Number:  
Price
371,00 €
Log in for availability
1 * 50 µG
Product Details & Documents

Produced in E. coli.

Specifications

Specifications

  • Pk:
    50 µG
  • Cat. No.:
    ENZOADISPP540050
  • Protein/peptide type:
    Recombinant
  • Purification:
    Purified by multi-step chromatography.
  • Protein/peptide name:
    Hsp 60
  • Formulation:
    Liquid. In Dulbecco's PBS.
  • Purity:
    >90% (SDS-PAGE; Western blot)
  • Species:
    Human
  • Molecular weight:
    ~60 kDa

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.