Order Entry
Germany
Orders LinkContactUsLinkComponent
Shoe covers

Shoe covers

PP non woven, White, 150 mm, 360 mm
Catalog # GMEQOJP012
Supplier:   GM EQUIPEMENT
Shoe covers
Catalog # GMEQOJP012
Supplier:  GM EQUIPEMENT
Shoe covers
Shoe covers

PP non woven, White, 150 mm, 360 mm

Price
51,70 €
Log in for availability
1 * 500 ST
Product Details & Documents

These shoe covers for laboratory use are available in various materials and sizes.

  • Elasticated opening for increased strength
  • Non slip sole (113-0678)
  • Waterproof (113-0677)

Documents

Specifications
  • Colour:
    White
  • Description:
    PP non woven
  • Height:
    150 mm
  • Length:
    360 mm
  • Seq:
    03
  • Cat. No.:
    GMEQOJP012
  • Garment application:
    Laboratory