MMP Red™ Substrate
MMP Red™ substrate, 1 mg
Catalog # AATB13521
Supplier: AAT Bioquest
Jump to:
Product Details & Documents
The matrix metalloproteinases (MMPs) constitute a family of zinc-dependent endopeptidases that function within the extracellular matrix.
- Bright signal
- Consistent results
- In-house quality testing
Caution: For research use only.
Specifications
Specifications
- Pk:1 mg
- Description:MMP Red™ substrate