Order Entry
Germany
Orders LinkContactUsLinkComponent
 

GAP Hazardous Material Work Stations

Hazardous material work station GAP.140.120.060, 1200×600×1400 mm, 1165×500×1105 mm, 80,70 kg
Catalog # MSGA40586901
Supplier:   asecos
GAP Hazardous Material Work Stations
Catalog # MSGA40586901
Supplier:  asecos
GAP Hazardous Material Work Stations

Hazardous material work station GAP.140.120.060, 1200×600×1400 mm, 1165×500×1105 mm, 80,70 kg

Price
6 330,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Hazardous material work station, GAP model with robust construction made of chemically resistant, anodised aluminium profiles.

  • Required occupational exposure limits are maintained
  • Avoidance of dangerous explosive atmosphere
  • Used for testing of the air equipment according to EN 14175 Part 3 (5.4.4)
  • Maximum user safety
  • Transparent side panes made from 5 mm (toughened safety glass)
  • Highly effective capturing of hazardous materials inside the work station through special fresh air curtain technology
  • Prepared for connection to the on-site exhaust air system and for electrical supply (230 V/50 Hz)
  • For mounting on existing working surfaces

Standard equipment consists of:
Melamine resin coated rear wall
Exhaust and fresh air monitoring with visual and audible alarm
Interior lighting aranged anti-glare (LED 4000 K neutral white 500 lux on the work surface)
Transparent side walls
Opaque, fold-away deflector
Potential-free switching contact 4-pole (max.400 V/25 A)
Power cord 3 m

Specifications
  • Weight:
    80,70 kg
  • W×D×H ext.:
    1200×600×1400 mm
  • Description:
    Hazardous material work station GAP.140.120.060
  • Seq:
    06
  • W×D×H int.:
    1165×500×1105 mm