GAP Hazardous Material Work Stations
Hazardous material work station GAP.110.090, 900×750×1100 mm, 865×650×855 mm, 103,20 kg
Catalog # MSGA40564901
Supplier: asecos
Jump to:
Product Details & Documents
Hazardous material work station, GAP model with robust construction made of chemically resistant, anodised aluminium profiles.
- Required occupational exposure limits are maintained
- Avoidance of dangerous explosive atmosphere
- Used for testing of the air equipment according to EN 14175 Part 3 (5.4.4)
- Maximum user safety
- Transparent side panes made from 5 mm (toughened safety glass)
- Highly effective capturing of hazardous materials inside the work station through special fresh air curtain technology
- Prepared for connection to the on-site exhaust air system and for electrical supply (230 V/50 Hz)
- For mounting on existing working surfaces
Standard equipment consists of:
Melamine resin coated rear wall
Exhaust and fresh air monitoring with visual and audible alarm
Interior lighting aranged anti-glare (LED 4000 K neutral white 500 lux on the work surface)
Transparent side walls
Opaque, fold-away deflector
Potential-free switching contact 4-pole (max.400 V/25 A)
Power cord 3 m
Specifications
- Weight:103,20 kg
- W×D×H ext.:900×750×1100 mm
- Description:Hazardous material work station GAP.110.090
- Seq:03
- W×D×H int.:865×650×855 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



