Orders LinkContactUsLinkComponent
Gel documentation system, Quantum CX5 Edge
Gel documentation system, Quantum CX5 Edge
Gel documentation system, Quantum CX5 Edge, with Epi UV, blue
Catalog # VILB101176791
Supplier:   Vilber Lourmat
Gel documentation system, Quantum CX5 Edge
Catalog # VILB101176791
Supplier:  Vilber Lourmat
Gel documentation system, Quantum CX5 Edge
Gel documentation system, Quantum CX5 Edge
Gel documentation system, Quantum CX5 Edge, with Epi UV, blue
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
The Quantum CX5 Edge gel documentation system is an ultra-modern, fully automated, yet compact system with innovative features and superior technology. With just one click, it allows the recording of images of the highest quality ('One-Button-Imaging').
  • Easy to operate, standalone
  • 'Scientific-grade' camera with 5 MP resolution and 6× automated zoom lens
  • Robust and light-proof all-metal darkroom
  • Including extendable transilluminator (optional as UV, SUPER-BRIGHT, blue and white light) usable with only one hand and interchangeable
  • Unprecedented sensitivity on a receiving surface of up to 21×26 cm
For maximum flexibility in fluorescence applications an optional Epi-UV illumination unit can also be installed. Thus the system can be perfectly adapted to experimental conditions.
 : The powerful BIO-VISION software (included) offers extensive possibilities of recording, display, analysis and output, and is clearly designed and easy to use.
Specifications
  • Description:
    Gel documentation system, Quantum CX5 Edge, with Epi UV, blue
  • Seq:
    03
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Power cord with EU plug
    Power cord with EU plug
    733-2833
    Log in for availability
    1 * 1 ST - 6,20 €