Masterflex® Single-Use Filling Needles
Product Details & Documents
A high-quality engineered solution for every application in the fill-finish workflow.
- Material compliant to USP Class VI, FDA 21 CFR, ADCF
- Double bagged in ISO Class 7 cleanroom
- High purity and contamination-free materials
- Ready to use; increases productivity
- Straightness <2 mm/M for consistent uniformity
- Oil-free and adhesive-free construction
- Repeatable performance with high-speed and accuracy
Filling speed, precision and accuracy are at the heart of the design process. Masterflex® Single Use Filling Needles are a cost‐effective and unique alternative to conventional re‐usable filling needles which require tedious and expensive sterilization and validation processes. They come in a variety of sizes engineered to work with Masterflex® L/S Peristaltic Pump Tubing ranging from L/S 14 to I/P 73 to facilitate a range of filling volumes. The Masterflex® Single Use Filling Needles are easily integrated into new or existing single‐use filling lines where the fluid path must be gamma irradiated or autoclaved. Products have a final cleaning and inspection in an ISO Class 7 cleanroom. All filling needles are cleaned with 70% IPA and vertically dried under HEPA-filtered air. Each needle is inspected before sealing in polybags inside the ISO cleanroom.
Documents
Certificates
Search by Lot NumberSpecifications
- Barb size/tubing int.Ø (port 1):¹⁄₁₆"
- Connection Type:Barbed Filling Needle
- Flow pattern:Straight
- Description:Single-use filling needle for Masterflex L/S 13 and L/S 14 peristaltic pump tubing; EA
- Seq:01
- Fitting material:Polycarbonate Housing, Stainless Steel Needle
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



