ContactUsLinkComponent
GW4869 CAS 6823-69-4 1x 5mg Storage (From the date of receipt):,, 3 years at -20C as powder 1 year at -80C in solvent 1 * 5 mg
Catalog # SELKS7609-5MG
Supplier:  SELLECK CHEMICALS
GW4869 CAS 6823-69-4 1x 5mg Storage (From the date of receipt):,, 3 years at -20C as powder 1 year at -80C in solvent 1 * 5 mg
Catalog # SELKS7609-5MG
Supplier:  SELLECK CHEMICALS