ContactUsLinkComponent
ERBB2/CEN 17 CISH Probe is designed for the qualitative detection of human ERBB2 gene amplifications as well as the detection of chr omosome 17 alpha satellites in formalin-fixed, paraffin-embedded specimens by chromogenic in situ hybridizatio 1 * 100 µl
Catalog # ABNOCG0033.100UL
Supplier:  Abnova
ERBB2/CEN 17 CISH Probe is designed for the qualitative detection of human ERBB2 gene amplifications as well as the detection of chr omosome 17 alpha satellites in formalin-fixed, paraffin-embedded specimens by chromogenic in situ hybridizatio 1 * 100 µl
Catalog # ABNOCG0033.100UL
Supplier:  Abnova