Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
HYDROMETER ISO649 S50 1.150-1.200:0.002 1 * 1 items
Catalog # ALLA3500SC0115/20
Supplier:
Alla France
Expand image
HYDROMETER ISO649 S50 1.150-1.200:0.002 1 * 1 items
Catalog # ALLA3500SC0115/20
Supplier:
Alla France