Orders LinkContactUsLinkComponent
Filter sets
Filter sets
Filter set 91 (high efficiency) reflector module F, excitation wavelengths 385, 475, 555 and 630 nm, consisting of filters TFT 450 + 538 + 610 and TBP 467/24 + 555/25 + 687/145
Catalog # ZEIS4890919110400
Supplier:   ZEISS
Filter sets
Catalog # ZEIS4890919110400
Supplier:  ZEISS
Filter sets
Filter set 91 (high efficiency) reflector module F, excitation wavelengths 385, 475, 555 and 630 nm, consisting of filters TFT 450 + 538 + 610 and TBP 467/24 + 555/25 + 687/145
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Filter set 91 (high efficiency) reflector module F, excitation wavelengths 385, 475, 555 and 630 nm, consisting of filters TFT 450 + 538 + 610 and TBP 467/24 + 555/25 + 687/145