Orders LinkContactUsLinkComponent
Filter sets
Filter sets
Filter set 90 (high efficiency) reflector module F, excitation wavelengths 385, 475, 555 and 630 nm, consisting of filters QBS 405 + 493 + 575 + 653 and QBP 425/30 + 514/30 + 592/25 + 709/100
Catalog # ZEIS4890909110400
Supplier:   ZEISS
Filter sets
Catalog # ZEIS4890909110400
Supplier:  ZEISS
Filter sets
Filter set 90 (high efficiency) reflector module F, excitation wavelengths 385, 475, 555 and 630 nm, consisting of filters QBS 405 + 493 + 575 + 653 and QBP 425/30 + 514/30 + 592/25 + 709/100
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Filter set 90 (high efficiency) reflector module F, excitation wavelengths 385, 475, 555 and 630 nm, consisting of filters QBS 405 + 493 + 575 + 653 and QBP 425/30 + 514/30 + 592/25 + 709/100