Orders LinkContactUsLinkComponent
Handheld Wireless Handle Barcode Scanner; Bluetooth, 1D, 2D, QR Code, CR2700
CR2700 scanner, light grey,handled version, kit with B27 battery, inductive charging station with desktop mount, and 1,8 m USB charging cable, W×D×H: 53,3×131,0×135,0 mm
  BRDY176687
 :   Brady
Handheld Wireless Handle Barcode Scanner; Bluetooth, 1D, 2D, QR Code, CR2700
  BRDY176687
 :  Brady
Handheld Wireless Handle Barcode Scanner; Bluetooth, 1D, 2D, QR Code, CR2700
Handheld Wireless Handle Barcode Scanner; Bluetooth, 1D, 2D, QR Code, CR2700
CR2700 scanner, light grey,handled version, kit with B27 battery, inductive charging station with desktop mount, and 1,8 m USB charging cable, W×D×H: 53,3×131,0×135,0 mm
1 170,00 €
1 * 1 ST
Easily read wide and small barcodes with the ultra-fast CR2700 Handled barcode scanner. This aggressive, omnidirectional scanner eliminates repositioning and time consuming re-scans of barcodes.
  • Uses B27 batteries with built-in power gauge
  • IP 65 seal rating against dust and liquid ingress
  • CodeShield® housing for disinfectant and chemical wipe down
  • Unequalled performance reading on curved and shiny surfaces
  • USB, Bluetooth®5 Low Energy (for enhanced data security, extended battery life, and less downtime)
Customisable visual, audible and haptic feedback with multiple programmable buttons. Trace any troubles with trackable service dates and battery statistics to help IT deploy and manage their scanner fleet.
  • Description:
    CR2700 scanner, light grey,handled version, kit with B27 battery, inductive charging station with desktop mount, and 1,8 m USB charging cable, W×D×H: 53,3×131,0×135,0 mm
  • Seq:
    03
Accessories
  •  
    Description
    For
  • BRDY145445
    Workstation lockout writer software app
    BBP®31, BBP®33, BBP®85, i3300
    1 * 1 ST - 833,00 €
  •  
    Description
    For
  • BRDY149445
    Brady workstation scan and print software suite
    BBP11, BBP12, BBP31, BBP33, BBP35, BBP37, BBP81, BBP85, BMP41, BMP51, BMP53, BMP61, BMP71, GlobalMark 2 Colour and Cut, GlobalMark 2 Multicolour, i3300, i5100, i7100, i7500, IDXPERT, IP Series, J2000, J4000, J5000, LABXPERT, M410, M510, M610, M611, M710, PR Plus, S3100, THT 3" Core Printers, TLS 2200
    1 * 1 ST - 833,00 €
  •  
    Description
  • BRDY195526
    Brady workstation CLP/GHS app via download
    1 * 1 ST - 369,99 €
  •  
    Description
    For
  • BRDY149187
    Brady workstation automation software suite
    BBP11, BBP12, BBP30, BBP33, BBP35, BBP37, BBP85, BMP41, BMP51, BMP53, BMP61, BMP71, GlobalMark 2 Multicolor, i3300, i5100, i7100, IP Series, J2000, J4000, J5000, M610, M611, M710, PR Plus, S3000, THT 76 mm Core Printers, Wraptor, Wraptor A6500
    1 * 1 ST - 833,00 €