Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ITS1 primer 25 nmole DNA Oligo Purification: Standard Desalting Sequence: TCC GTA GGT GAA CCT GCG G Guarantee 12 nmol 1 * 1 ST
Catalog # IDNAITS1
Supplier:
INTEGRATED DNA TECHNOLOGIE
Expand image
ITS1 primer 25 nmole DNA Oligo Purification: Standard Desalting Sequence: TCC GTA GGT GAA CCT GCG G Guarantee 12 nmol 1 * 1 ST
Catalog # IDNAITS1
Supplier:
INTEGRATED DNA TECHNOLOGIE