S-Trap micro sample processing in a full kit format for samples = 100 µg. The kit includes S-Trap micros, extra tubes, and lysis, wash and digestion buffers. The user must supply HPLC or MS grade MeOH and their choice of protease. 160 Isolations 1 * 160 ST
Catalog # MSPPK02-MICRO-160
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together