Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
1 component: 1,2,3-Trimethylbenzene [CAS:526-73-8] 2000mg/l in Methanol 1 * 5 Ampoul
Catalog # CMPLRD0533051.5
Supplier:
CUSTOM MADE CHEMICALS LAB
Expand image
1 component: 1,2,3-Trimethylbenzene [CAS:526-73-8] 2000mg/l in Methanol 1 * 5 Ampoul
Catalog # CMPLRD0533051.5
Supplier:
CUSTOM MADE CHEMICALS LAB