Orders LinkContactUsLinkComponent
Masterflex® Sanitary Tees | Avantor
Masterflex® Sanitary Tees
Fitting, Sanitary Tee, 1½", Polypropylene; EA, Sanitary, Polypropylene, 1½"
Catalog # MFLX40618-80
Masterflex® Sanitary Tees
Catalog # MFLX40618-80
Masterflex® Sanitary Tees
Masterflex® Sanitary Tees
Fitting, Sanitary Tee, 1½", Polypropylene; EA, Sanitary, Polypropylene, 1½"
Wechseln zu:
Price
112,00 €
Anmelden für die Verfügbarkeit
1 * 1 ST
Product Details & Documents
These compliant, biocompatible fittings provide assurance, integrity, and cost-effective solutions.
  • Engineered to meet the needs of bioprocess workflows, single-use tees uphold the highest standards of compliance to meet critical needs
  • Created for flawless sterility, sanitary tees can undergo gamma irradiation and autoclaving, ensuring decontamination in all processes
  • Constructed from USP Class VI materials, sanitary tees prioritise biocompatibility and safety, guaranteeing their suitability for sensitive bioprocess applications
  • Designed for easy integration, single-use tees are engineered to provide reliable connections and consistent performance
Sanitary single-use tees are engineered and manufactured from gamma irradiatable and autoclavable Polypropylene and PVDF USP Class VI materials. These fittings offer an uncompromising commitment to sterilisation. Manufactured within an ISO Class 7 cleanroom, these sanitary tees provide the highest level of biocompatibility and safety, ensuring seamless integration into critical processes within the biopharmaceutical industry.
Spezifikationen:
  • Connection Type:
    Sanitary
  • Sanitary clamp size (Port 1):
    1½"
  • Description:
    Fitting, Sanitary Tee, 1½", Polypropylene; EA
  • Seq:
    03
  • Fitting material:
    Polypropylene
  • Cat. No.:
    MFLX40618-80
  • Sterilisation:
    Gamma, Autoclave
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.