Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.002 mg, max. load 111 g. Draft Shield D 1 * 1 items
SARBMCA116S3CE3
:
Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.002 mg, max. load 111 g. Draft Shield D 1 * 1 items
SARBMCA116S3CE3
:
Sartorius Balances