Calibrated solution used to allow user to verify correct wavelength and photometric performance of the instrument 1 * 1 ST
Catalog # NANDCHEM-PV-8
Supplier: THERMO NANODROP
Calibrated solution used to allow user to verify correct wavelength and photometric performance of the instrument 1 * 1 ST
Catalog # NANDCHEM-PV-8
Supplier: THERMO NANODROP
Frequently Bought Together
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
Ethylenediamine 99%
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
2,2,4-Trimethylpentane 99.5% for analysis
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
Supplier: Thermo Fisher Scientific
Toluene ≥98%, TECHNICAL
Supplier: VWR Chemicals
2,2,4-Trimethylpentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanol, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetone ≥99.5% (by GC), liquid USP, BP, Ph. Eur., pure
Supplier: PanReac AppliChem
n-Octane, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Bromoform 99+% stabilised
Supplier: Thermo Fisher Scientific
HYDROXYLAMINE HCl 1 % 1 * 10 L
Supplier: Bernd Kraft