ADC Active Digital Controller - LED Digital Display (mbar/Torr/Pa/V) supports 1 Gauge Head (from APG APGX & WRG) - Power Supply 100-240V ac 47-63 Hz 1Ø IEC320 Plug - Dimensions DIN43700 90mm x 43mm x 159mm Deep 1 * 1 ST
Catalog # EDWAD39590000
Supplier: EDWARDS HIGH VACUUM METTLER