Skydrol 500B-4 BMS3-11P Type IV Class II 1 * 0,95 L
MSPPS500B40USQ
Frequently Bought Together
ADF Volumetric Solution for analysis of animal feed (NF V18-122)
: VWR Chemicals
TENOFOVIR ALAFENAMIDE HEMIFUMARATE 10MM (IN 1ML DMSO
: MARKET SOURCE PART PROCESS
[EN]IGG4 ISOTYPE CONTROL SPLE/R409K 1 * 1 mg
: MARKET SOURCE PART PROCESS
[EN]FILMOLUX 609 50M X 10CM 40 MM KERN 1 * 1 Rolle
: MARKET SOURCE PART PROCESS
Methyl green zinc chloride salt <0,5% crystal violet 80% (dye content), Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
Acid Phosphatase, Leukocyte (TRAP) Kit Select reagents packaged in gelatin capsules, Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
Test cassettes for urine
: SURESCREEN