ContactUsLinkComponent
L-MARK Gold Syringe 100UL Fixed Needle, Black PTFE, 51mm Length, 22s Gauge, LC Tip PK1 1 * 1 items
Catalog # SGEALMK.2620719CBL
L-MARK Gold Syringe 100UL Fixed Needle, Black PTFE, 51mm Length, 22s Gauge, LC Tip PK1 1 * 1 items
Catalog # SGEALMK.2620719CBL
Frequently Bought Together