L-MARK Syringe 5ML Remov Needle, PTFE, 57mm Length, 22 Gauge, LC Tip PK1 1 * 1 items
Catalog # SGEALMK.3030097
Supplier: Trajan Scientific and Medical
L-MARK Syringe 5ML Remov Needle, PTFE, 57mm Length, 22 Gauge, LC Tip PK1 1 * 1 items
Catalog # SGEALMK.3030097
Supplier: Trajan Scientific and Medical
Frequently Bought Together
Tubing connectors, b.safe
Supplier: Bohlender
Safety waste caps and ventilation kit, SymLine®
Supplier: S.C.A.T.
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Supplier: VWR Collection
VWR®, Screw Neck Vials ND15
Supplier: VWR Collection
Sample vials, E-C, WHEATON®
Supplier: DWK Life Sciences
QuEChERS mixes, clean-up, CHROMABOND®
Supplier: MACHEREY-NAGEL
Capillary column ferrules for gas chromatography
Supplier: Agilent
Accessories for 7500/7700
Supplier: Agilent
γ-BHC (lindane)
Supplier: Restek
Immobilised polysaccharide columns
Supplier: CHIRAL TECHNOLOGIES
Spare needles for autosampler syringes
Supplier: Agilent
Gold standard syringes for autosampler
Supplier: Agilent
Avantor® ACE® C8-300, HPLC Columns, 3 µm
Supplier: Avantor
TSKgel® gel filtration columns, PW-type
Supplier: TOSOH Bioscience
GC columns, Ultra Inert, J&W DB-WAX
Supplier: Agilent