N-[9-[(6aR,8R,9R,9aS)-9-hydroxy-2,2,4,4-tetraisopropyl-6a,8,9,9a-tetrahydro-6H-furo[3,2-f][1,3,5,2,4]trioxadisilocin-8-yl]-6-oxo-3H- purin-2-yl]-2-methyl-propanamide 1 * 1 g
Catalog # PHBLPBQS505-1G
Supplier: Pharmablock (USA)
N-[9-[(6aR,8R,9R,9aS)-9-hydroxy-2,2,4,4-tetraisopropyl-6a,8,9,9a-tetrahydro-6H-furo[3,2-f][1,3,5,2,4]trioxadisilocin-8-yl]-6-oxo-3H- purin-2-yl]-2-methyl-propanamide 1 * 1 g
Catalog # PHBLPBQS505-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
1-Butanol ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol eurodenatured ≥99%, TechniSolv® (1:1:1)
Supplier: VWR Chemicals
Cyclohexane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals