ANTI-SARS-COV-2 SPIKE RBD ANTIBODY MOUSe IgG1 (9A6A11) (XBB.1.5/Omicron Specific) (MALS verified) 1 * 1 mg
Catalog # ACRBSPD-S298-1MG
Supplier: ACRO Biosystems
ANTI-SARS-COV-2 SPIKE RBD ANTIBODY MOUSe IgG1 (9A6A11) (XBB.1.5/Omicron Specific) (MALS verified) 1 * 1 mg
Catalog # ACRBSPD-S298-1MG
Supplier: ACRO Biosystems
Frequently Bought Together
Microplates, 96-well
Supplier: BOETTGER PAUL
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR®, Cell Culture Flasks, Non Treated or Treated
Supplier: VWR Collection
Sampling scoops, SteriPlast®
Supplier: BURKLE
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
VWR® Media Bottles, Square, PETG
Supplier: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
Petri dishes, Corning® Gosselin™
Supplier: Corning
J.T.Baker®, Plate Medio
Supplier: Avantor
VWR®, Flat Screw Caps for Microcentrifuge Tubes
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
Corning® CoolCell® containers
Supplier: Corning
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
Petri dishes
Supplier: Greiner Bio-One
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
Storage bottles, octogonal, PET, Corning® Gosselin™
Supplier: Corning
Bottles, narrow neck, octagonal, PET, with screw caps
Supplier: SEMADENI
Microcentrifuge tubes, 0,5/0,7/1,5/2,0/2,2 ml
Supplier: Greiner Bio-One