ContactUsLinkComponent
Wire / Cable Scrape Abrasion Kit Test the durability of wire insulation in accordance with ISO 6722. The kit is used with the Taber Linear Abraser model 5750 after s/n 20091391, and should be mounted to either the T-Slot Universal Table (p/n 133336) 1 * 1 KIT
Catalog # SIMETB0000-134887
Supplier:  Industrial Physics
Wire / Cable Scrape Abrasion Kit Test the durability of wire insulation in accordance with ISO 6722. The kit is used with the Taber Linear Abraser model 5750 after s/n 20091391, and should be mounted to either the T-Slot Universal Table (p/n 133336) 1 * 1 KIT
Catalog # SIMETB0000-134887
Supplier:  Industrial Physics