Orders LinkContactUsLinkComponent
pH Sensor, Memosens CPS61E
pH Sensor, Memosens CPS61E
Memosens CPS61E pH sensor, 225 mm, 0 - 14 pH, 0...100 °C, 0,8...7 bar (abs), sterilisable up to 140 °C; EC_00831_03.20; Nonhazardous area approval
Catalog # 662-2463
Supplier:   Endress+Hauser
pH Sensor, Memosens CPS61E
Catalog # 662-2463
Supplier:  Endress+Hauser
pH Sensor, Memosens CPS61E
pH Sensor, Memosens CPS61E
Memosens CPS61E pH sensor, 225 mm, 0 - 14 pH, 0...100 °C, 0,8...7 bar (abs), sterilisable up to 140 °C; EC_00831_03.20; Nonhazardous area approval
Price
927,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Memosens 2.0 glass sensor for the food and beverage industry and for bioreactors in biotech industry.
  • Memosens 2.0 offers extended storage of calibration and process data, enabling better trend identification and providing a future-proof basis for predictive maintenance and enhanced IIoT services
  • The sensor's exceptional accuracy and reproducibility help you to keep the pH value in the optimum range for a maximized product yield
  • A special glass membrane and reference system makes the sensor CIP/SIP and autoclaving resistant (up to 140 °C/284 °F) offering maximum long-term stability
  • The sensor protects your product quality with its certified biocompatibility according to USP class VI, FDA compliance of wetted parts, VO 1935/2004 compliance, no cytotoxicity, bioreactivity or animal-based materials
  • Maximum process safety through non-contact, inductive signal transmission
  • Reduced operating costs thanks to minimized process downtime and extended sensor lifetime
Keep the pH value in the optimum range and maximise your product yield with Memosens CPS61E. Its rugged design and long-term stability ensure extremely accurate and reproducible measured values even after CIP/SIP or autoclaving. CPS61E features Memosens 2.0 digital technology, offering extended storage of calibration and process data for predictive maintenance. The sensor resists moisture and enables pre-calibration in the laboratory. This enhances the process integrity and increases the process uptime.
Memosens CPS61E measures pH in: Biotech and pharma industries; bioreactors/fermenters: process control in enzyme production; bioreactors/fermenters: control of culture growth; food and beverage; CIP/SIP applications.
A pharma certificate of compliance with sensor serial number specified is available.
Specifications
  • Description:
    Memosens CPS61E pH sensor, 225 mm, 0 - 14 pH, 0...100 °C, 0,8...7 bar (abs), sterilisable up to 140 °C; EC_00831_03.20; Nonhazardous area approval
  • Cat. No.:
    662-2463
  • Sensor type:
    Hygienic version, zero point pH 7,0, temperature sensor NTC 30k
  • Reference system:
    Pressurised, 1 ceramic junction, ion trap, 3 M KCl, Ag/AgCl
  • Range:
    0 - 14 pH, 0...100 °C, 0,8...7 bar (abs) , sterilisable up to 140 °C (application range)