ABSCISIC ACID ELISA QUANTITATION KIT 1 * 96 Tests
Catalog # AGRIAS204392
Supplier: Agrisera
Frequently Bought Together
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
VWR®, Centrifuge Tubes, Conical Bottom, with Flat Screw Cap
Supplier: VWR Collection
Test tubes
Supplier: Greiner Bio-One
Transfer pipettes
Supplier: RATIOLAB
VWR® Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection
Cell culture flasks, Nunc™
Supplier: Thermo Fisher Scientific
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Round-bottom polystyrene tubes, disposable, Falcon®
Supplier: Corning
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
Adhesive PCR film and foil seals
Supplier: Thermo Fisher Scientific
Centrifuge tubes, 15 ml, Corning®
Supplier: Corning
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR®, Petri Dish, PS
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR® Water Sampling Bottles, with Tamper Evident Cap
Supplier: VWR Collection