Order Entry
Germany
Orders LinkContactUsLinkComponent
 

SkillPak™ BIO Pre-Packed Columns

Method development kit, Protein L, PP, 1000 Å, 45 µm, 16,00 mm, 25 mm, 5 ml, TOYOPEARL AF-rProtein L-650F
Catalog # TOYO0045360
Supplier:   TOSOH Bioscience
SkillPak™ BIO Pre-Packed Columns
Catalog # TOYO0045360
Supplier:  TOSOH Bioscience
SkillPak™ BIO Pre-Packed Columns
SkillPak™ BIO Pre-Packed Columns

Method development kit, Protein L, PP, 1000 Å, 45 µm, 16,00 mm, 25 mm, 5 ml, TOYOPEARL AF-rProtein L-650F

Price
5 940,00 €
Log in for availability
1 * 6 ST
Product Details & Documents

SkillPak™ BIO pre-packed columns are designed for a fast method development and scale-up of multi-column chromatography (MCC) or continuous chromatography processes. They are pre-packed with TOYOPEARL® process chromatography media for bioseparations.

  • MCC-dedicated dimensions and packing procedures
  • Comprehensive resins for MCC applications
  • Multi-column kits with single resin lot and low column-to-column variability

They are ready to use and show excellent physical strength and ideal pressure-flow characteristic for downstrem processing in multi-column (MCC) or continuous (CC) operations. Pre-packed with TOYOPEARL®process chromatography media, these columns offer superior separation of biomolecules, such as monoclonal antibodies. SkillPak™ BIO columns are available in multi-column kits packed with a single resin lot and low column-to-column variability to ensure low comparability between columns in MCC and CC.

All SkillPak BIO columns guarantee optimal performance and can be operated on all MCC and CC systems, as well as standard low-or medium-pressure liquid chromatography systems. They are reproducibly packed and take into account the varying compressibility of each resin.

Specifications
  • Column Format:
    Method development kit
  • Ø int.:
    16,00 mm
  • Particle Size:
    45 µm
  • Pore size:
    1000 Å
  • Capacity:
    5 ml
  • Length:
    25 mm
  • Seq:
    11
  • Column hardware:
    PP
  • Phase:
    Protein L
  • Packing material:
    TOYOPEARL AF-rProtein L-650F