Sep-Pak Alumina A 20 cc Vac Cartridge, 5 g Sorbent per Cartridge, 50 - 300 µm 1 * 20 items
Catalog # WATEWAT054670
Supplier: WATERS
Sep-Pak Alumina A 20 cc Vac Cartridge, 5 g Sorbent per Cartridge, 50 - 300 µm 1 * 20 items
Catalog # WATEWAT054670
Supplier: WATERS
Frequently Bought Together
20Mm Borate Buffer Ph9.3 100Ml 1 * 100 mL
Supplier: Agilent
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
ISAspher HPLC Columns, Silica
Supplier: ISERA
Coated polysaccharide columns, reversed phase
Supplier: CHIRAL TECHNOLOGIES
Headspace crimp vials, 20 mm
Supplier: Restek
LC vials and closures
Supplier: Agilent
XBridge BEH C8 Column, 130Å, 5 µm, 4.6 mm X 50 mm 1 * 1 items
Supplier: WATERS
Acrylamide Starter Kit UHPLC EnhClnUp 1 * 1 KIT
Supplier: WATERS
Screw caps for screw neck vials, N 9
Supplier: MACHEREY-NAGEL
Accessories
Supplier: Bohlender
PROTEINWORKS EXPRESS DIGEST START-UP KIT 1 * 1 KIT
Supplier: WATERS
Plugs, PE
Supplier: MACHEREY-NAGEL
Disposable tip adapters
Supplier: HAMILTON BONADUZ