CORTECS T3 Method Validation Kit, 120Å, 2.7 µm, 3 mm X 150 mm 1 * 3 items
Catalog # WATE186008523
Supplier: WATERS
CORTECS T3 Method Validation Kit, 120Å, 2.7 µm, 3 mm X 150 mm 1 * 3 items
Catalog # WATE186008523
Supplier: WATERS
Frequently Bought Together
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Avantor® ACE® C18, HPLC Columns, 5 µm
Supplier: Avantor
Spillage trays
Supplier: S.C.A.T.
SCREW CAP PHENOLIC 15MM BUTYL/PTFE 1 * 100 items
Supplier: DWK Life Sciences
Glass crimp top headspace vials, 20 ml, level 2 high-throughput applications, SureSTART™
Supplier: Thermo Fisher Scientific
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Supplier: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
VWR®, Screw Neck Vials ND10
Supplier: VWR Collection
VWR®, Snap Ring Vials ND11
Supplier: VWR Collection
20Mm Borate Buffer Ph9.3 100Ml 1 * 100 mL
Supplier: Agilent
FlashPure EcoFlex Silica Cartridges
Supplier: BUCHI
Safety canisters, electrically conducting
Supplier: S.C.A.T.
SUPELCOSIL LC-SI 5UM 15CMX4.6MM HPLC COL UMN 1 * 1 items
Supplier: Merck
Manual microliter syringes, Hamilton 700 series
Supplier: Restek
Avantor® ACE® Excel® NH₂, HPLC/UHPLC Columns, Analytical, 1,7 µm
Supplier: Avantor