XSelect CSH Phenyl-Hexyl Method Validation Kit, 130Å, 3.5 µm, 3 mm X 100 mm 1 * 3 items
WATE186005561
: WATERS
XSelect CSH Phenyl-Hexyl Method Validation Kit, 130Å, 3.5 µm, 3 mm X 100 mm 1 * 3 items
WATE186005561
: WATERS
Frequently Bought Together
Accessories for sample tube
: METROHM
Avantor® ACE® C18, HPLC Columns, 5 µm
: Avantor
250R-GT 250UL SYRINGE CALIBRATED 1 * 1 items
: Trajan Scientific and Medical
Capillary columns, PERMABOND® Silane
: MACHEREY-NAGEL
Avantor® Alltima, HPLC Columns, C18
: Avantor
Chromatography Tubing, Stainless Steel
: Avantor
Specification certified screw vial and cap kits, level 2 high-throughput applications, SureSTART™
: Thermo Fisher Scientific
Electronic hand-held crimpers and decrimpers, level 2 high throughput applications, SureSTART™
: Thermo Fisher Scientific