Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
XSelect HSS Cyano (CN) Method Validation Kit, 100Å, 3.5 µm, 3 mm X 150 mm 1 * 3 items
Catalog # WATE186005952
Supplier:
WATERS
Expand image
XSelect HSS Cyano (CN) Method Validation Kit, 100Å, 3.5 µm, 3 mm X 150 mm 1 * 3 items
Catalog # WATE186005952
Supplier:
WATERS