ACQUITY UPLC HSS C18 Column, 100Å, 1.8 µm, 3 mm X 150 mm 1 * 3 items
Catalog # WATE176001820
Supplier: WATERS
ACQUITY UPLC HSS C18 Column, 100Å, 1.8 µm, 3 mm X 150 mm 1 * 3 items
Catalog # WATE176001820
Supplier: WATERS
Frequently Bought Together
20Mm Borate Buffer Ph9.3 100Ml 1 * 100 mL
Supplier: Agilent
Avantor® ACE® C18, HPLC Columns, 5 µm
Supplier: Avantor
FlashPure EcoFlex Silica Cartridges
Supplier: BUCHI
Septa for open top caps, WHEATON®
Supplier: DWK Life Sciences
HPLC columns, Hypersil™ BDS C8
Supplier: Thermo Fisher Scientific
Accessories for SCAT Safety Caps
Supplier: S.C.A.T.
Capillary columns, traditional phases, SPB-5, Supelco®
Supplier: Merck
Transition fittings, BOLA
Supplier: Bohlender
LC vials and closures
Supplier: Agilent
Gastight® 1000 series syringes
Supplier: HAMILTON BONADUZ
Tubing, PTFE
Supplier: HAMILTON BONADUZ
'Plunger in needle' syringes, 0,5 to 5 µl
Supplier: Trajan Scientific and Medical
HSPgel Column, 5 µm, 6 mm X 150 mm, 2K - 60K 1 * 1 items
Supplier: WATERS
Avantor® Partisil SAX, HPLC Columns
Supplier: Avantor